Learn More
Thermo Scientific™ BIOLiner™ Rotor Bucket Adapters
Combine Thermo Scientific™ BIOLiner™ Rotor Bucket Adapters with the BIOLiner Rotor Bucket for a complete system.
Manufacturer:  Thermo Scientific™ 75003673
 View more versions of this product
Catalog No. 75-003-673
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications
Description
Combine Thermo Scientific™ BIOLiner™ Rotor Bucket Adapters with the BIOLiner Rotor Bucket for a complete system.
Ordering Information:
Individual adapters and bottle/adapter packages are offered.
  • Capacity range: 3L to 980mL
  • Maximum tube dimensions (O.D. x L): 98 x 124mm
Includes:
75-003-692 includes 750mL flat-bottom bottle with all round adapters for the TX-750 round bucket and four 500mL Corning™ conical bottle adapters.
Description & Specifications
Specifications
15mL Conical Tubes
4