Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ 16,16-Dimethyl Prostaglandin E2
GREENER_CHOICE

Catalog No. 40271
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-271 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 40-271 Supplier Tocris Bioscience™ Supplier No. 4027/1
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Synthetic prostaglandin E2 (Cat. No. 2296) derivative; regulates HSC development

Synthetic derivative of prostaglandin E2 (Cat. No. 2296). Increases embryonic stem cell (ESC) hematopoietic colony formation in mouse bone marrow. Also induces an increase in hematopoietic stem cell (HSC) numbers and enhances Wnt activity within the HSC population in zebrafish embryos.

Chemical Identifiers

CAS 39746-25-3
Molecular Formula C22H36O5

Specifications

Quantity 1 mg
Formula Weight 380.52
Purity >98%
Molecular Weight (g/mol) 380.525
Chemical Name or Material 16, 16-Dimethyl Prostaglandin E2
Recommended Storage Store at -80°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.