Learn More
FUNGI PERFECTI 16 INCH IMPULSE SEALER
Manufacturer:  FUNGI PERFECTI ETIS16
Encompass
An excellent sealer, well-suited for use with sterile airflow spawn bags. Provides a seal up to 16 inches wide in approximately 1 second. This unit is an essential tool for the airtight sealing of bags used in the manufacture of sawdust blocks.
Catalog No. NC1645004