Learn More
17-Beta-Estradiol Mouse anti-All Species, Alexa Fluor™ 594, Clone: 4S112 (BGN/06/88112), Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB10062279AF594
Catalog No. N1062279A94
Add to cart
Description
Description
17-Beta-Estradiol Monoclonal antibody specifically detects 17-Beta-Estradiol in All Species samples. It is validated for ELISA.
Specifications
Specifications
17-Beta-Estradiol
4S112 (BGN/06/88112)
ELISA
17 beta Oestradiol
17-beta estradiol conjugated to bovine serum albumin at position 6.
0.1 mL
Primary
NB100-62279 is specific for 17-beta estradiol, a mammalian estrogenic hormone, produced in the ovaries, placenta and testis that has a role in normal sexual development and the regulation of the menstrual cycle. NB100-62279 exhibits negligible cross reactivity with estriol and testosterone.
ELISA
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
All