Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
17 beta-HSD14/HSD17B14 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$152.22 - $436.00
Specifications
Antigen | 17 beta-HSD14/HSD17B14 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15762520
|
Novus Biologicals
NBP15762520UL |
20 μL |
Each for $152.22
|
|
NBP157625
|
Novus Biologicals
NBP157625 |
100 μL |
Each for $436.00
|
|
Description
17 beta-HSD14/HSD17B14 Polyclonal specifically detects 17 beta-HSD14/HSD17B14 in Human samples. It is validated for Western Blot.Specifications
17 beta-HSD14/HSD17B14 | |
Polyclonal | |
Rabbit | |
Lipid and Metabolism | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
dehydrogenase/reductase (SDR family) member 10, Dehydrogenase/reductase SDR family member 10, DHRS1017-beta-hydroxysteroid dehydrogenase 14, EC 1.1.1.-, hydroxysteroid (17-beta) dehydrogenase 14,17-beta-hydroxysteroid dehydrogenase DHRS10, retinal short-chain dehydrogenase/reductase 3, Retinal short-chain dehydrogenase/reductase retSDR3, retSDR3, RETSDR3,17-beta-HSD 14, SDR3, SDR47C1, short chain dehydrogenase/reductase family 47C, member 1 | |
HSD17B14 | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
Q9BPX1 | |
51171 | |
Synthetic peptides corresponding to HSD17B14(hydroxysteroid (17-beta) dehydrogenase 14) The peptide sequence was selected from the middle region of HSD17B14. Peptide sequence QPAEVGAAAVFLASEANFCTGIELLVTGGAELGYGCKASRSTPVDAPDIP. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title