Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
17-PA, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective antagonist of neurosteroid potentiation and direct gating of GABAA receptors. Selectively reduces the effects of 5α-reduced steroids compared to 5β-reduced steroids and displays no effect on potentiation evoked by barbiturates and benzodiazepines. Attenuates 3α,5α-THP-induced loss of righting reflex and total sleep time following i.c.v administration in rats.
Specifications
Quantity | 10 mg |
Formula Weight | 350.54 |
Percent Purity | >99% |
Chemical Name or Material | 17-PA |
Safety and Handling
Recommended Storage : Store at Room Temperature