Learn More
Propper Verline™ Autoclave Indicator Tape
SDP
Verline™ Beige Autoclave Indicator Tape is a pressure-sensitive sterilization tape used to distinguish between processed and unprocessed items.
Supplier:  Propper 26814516
 View more versions of this product
Catalog No. 17-000-031
Add to Cart
Description
Description
  • Verline™ is a beige autoclave indicator tape used in steam sterilizers.
  • Featuring lead-free indicator ink, placed in green stripes that turn black after exposure to a steam sterilization cycle.
  • Verline™ tape's strong adhesive and even coating allow for even dispensing and easy application.
  • Specifically formulated universal adhesive provides secure bonding with fabric and non-woven wraps, paper, metal, and glass yet provide easy removal without residue traces.
  • AAMI/ISO 11140-1 Type 1 Compliant Process Indicator Tape
  • Not Made with Natural Rubber Latex
  • Steri-Wrap™ Unite™ Bonded SMS Wrap for Steam, EO and Plasma Sterilization
Specifications
Specifications
AAMI/ISO 11140-1 Type 1 Compliant
Steam Autoclave Tape
256/Cs.
14 yd.
19 mm
Beige
Yes
42 in.
0.75 in.