Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
High-Tech Conversions Tacky Traxx™ Entrance Mats
Tacky Traxx™ Entrance Mats are disposable sticky entrance mats for cleanroom, industrial/construction, and maintenance.
Supplier: High-Tech Conversions TT264534B
Description
- Numbered corner tabs for easy tracking of remaining sheets.
- Smooth, effective surface is not damaged by shoes, booties, or cart wheels.
- Peel-off sheets eliminate the need for messy cleaning and make it easy to maintain a clean surface.
- Perfect for cleanrooms, laboratories, construction, manufacturing, maintenance/installation service, and residential use.
Specifications
Cleanroom Tacky Mat | |
Perfect for cleanrooms, laboratories and any other particle restrictive environments, For entrance to or within particle-controlled environment, Eliminates messy cleaning | |
Polyethylene Film | |
46 in. | |
116.84 cm |
Yes | |
Blue | |
68.58 cm | |
27 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction