Thermo Scientific™ Storage Vials & Closures Available on GSA/VA Contract for Federal Government customers only.
Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample. Storage vials range from 2mL to 40mL.
Manufacturer: Thermo Scientific™ 18SCST
 View more versions of this product
Catalog No. 10-800-149
  Check Availability
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    Material Glass (Vial),Polypropylene (Cap),PTFE/Foam (Septum)
    Cap Style Screw Cap with Septum
    Closure Material PTFE Lined
    Vial Closure Size 18-400mm
    Thickness (Metric) 1.0mm
    Type Screw Cap
    Closure Color White
    Item Description 18-400 Screw Cap
    For Use With Storage Vials
    Quantity 100
    Septum PTFE/PE Foam Liner
    Description 18-400 Thread
    Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample. Thermo Scientific™ National™ and Thermo Scientific™ Chromacol™ Sample Storage Screw Thread Vials are manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) or 51A amber (Type 1, Class B) glass. Storage vials range from 2mL to 40mL.
    • PTFE-lined solid-top storage caps
    • PTFE on film/foam backing offers broad chemical resistance
    • Not autoclavable
    Order as individual sample storage vials and closures or as unassembled convenience kits.
    Unassembled convenience kits saves time during sample preparation.
    • Solid-top convenience kits include matched quantities of vials and screw caps with pre-assembled septa.
    • Open-top convenience kits contain shrink-wrapped vials and separately packaged caps and septa in polybags.
  • Description & Specifications
Related Products
Product Content Feedback