Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

18-Crown-6, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
500g
Packaging:
Glass bottle
17455-13-9
C12H24O6
264.32
XEZNGIUYQVAUSS-UHFFFAOYSA-N
18-crown-6,18-crown-6 ether,ethylene oxide cyclic hexamer,18-crown-6-ether,unii-63j177nc5b,ccris 3587,18-crown 6-ether,1,4,7,10,13,16-hexanoxacyclooctadecane,18-crown-6/1,4,7,10,13,16-hexaoxacyclooctadecane
28557
CHEBI:32397
1,4,7,10,13,16-hexaoxacyclooctadecane
C1COCCOCCOCCOCCOCCO1
17455-13-9
C12H24O6
264.32
XEZNGIUYQVAUSS-UHFFFAOYSA-N
18-crown-6,18-crown-6 ether,ethylene oxide cyclic hexamer,18-crown-6-ether,unii-63j177nc5b,ccris 3587,18-crown 6-ether,1,4,7,10,13,16-hexanoxacyclooctadecane,18-crown-6/1,4,7,10,13,16-hexaoxacyclooctadecane
28557
CHEBI:32397
1,4,7,10,13,16-hexaoxacyclooctadecane
C1COCCOCCOCCOCCOCCO1
Chelation/Complexation reagent
Packaging Glass bottle
Melting Point 36.0°C to 40.0°C
Color Colorless or White
Flash Point >110°C
Quantity 100g
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Fieser 04,142; 05,152; 06,133; 07,76; 08,1280; 09,126; 12,403; 15,76; 17,290
Merck Index 14,2602
Solubility Information Solubility in water: soluble. Other solubilities: soluble in hydrocarbons,benzene,chloroform,methanol and trichlorotrifluoroethane
Formula Weight 264.32
Physical Form Crystals or Crystalline Mass or Liquid
Percent Purity 99%
Chemical Name or Material 18-Crown-6
Show More Show Less

GHS Signal Word: Warning

EINECSNumber : 241-473-5