Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety 18 in. Orange Aramid Fiber Sleeve

18 in. Orange Aramid Fiber Sleeve features a patent-protected, high visibility orange aramid fiber yarn. The high visibility orange enhances safety awareness.

$40.14

Specifications

Color Orange
For Use With (Application) Assembly Line Production
Material Dupont™ Kevlar™ aramid fibers
Size One Size
Sterility Non-Sterile
View More Specs

Products 2
Catalog Number Mfr. No. Construction Price Quantity & Availability  
Catalog Number Mfr. No. Construction Price Quantity & Availability  
19-812-642 View Documents MCR Safety
9178OT
Thumb Slot Each for $40.14
Add to cart
 
19-812-643 View Documents MCR Safety
9178O
No Thumb Each for $40.14
Add to cart
 
Description

Description

  • Patent-protected, high visibility orange DuPont™ Kevlar™ aramid fibers® engineered yarn
  • 13 gauge seamless knit
  • Gussets for expandability

Specifications

Specifications

Orange
Dupont™ Kevlar™ aramid fibers
Non-Sterile
Assembly Line Production
One Size
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit