Alfa Aesar™ 18-Pentatriacontanone, 97+%

Antiblocking agent

Manufacturer: Alfa Aesar™ 4144514

Quantity 25g

 View more versions of this product

Catalog No. AA4144514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 504-53-0
    Linear Formula (CH3(CH2)16)2CO
    Formula Weight 506.94
    MDL Number MFCD00048491
    Chemical Name or Material 18-Pentatriacontanone

    Specifications

    Formula Weight 506.94
    MDL Number MFCD00048491
    Quantity 25g
    TSCA TSCA
    Physical Form Powder
    Solubility Information Soluble in benzene
    Color Off-white
    Melting Point 79° to 80°C
    Linear Formula (CH3(CH2)16)2CO
    CAS 504-53-0
    Chemical Name or Material 18-Pentatriacontanone
    Assay ≥97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit