Learn More
Tocris Bioscience™ 18A
HIV cell entry blocker
$145.00 - $609.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
561210 View Documents Tocris Bioscience™
5612/10
Each for $145.00
Add to cart
561250 View Documents Tocris Bioscience™
5612/50
Each for $609.00
Add to cart
Description
Description
HIV cell entry blocker. Binds gp120 and inhibits cell entry for a broad spectrum of HIV isolates. Exhibits no effect on the binding of CD4 or CCR5.