Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

18beta-Glycyrrhetinic Acid, Approx. 98%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
471-53-4
C30H46O4
470.69
MPDGHEJMBKOTSU-YKLVYJNSSA-N
(2S,4aS,6aS,6bR,8aR,10S,12aS,12bR,14bR)-10-hydroxy-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-icosahydropicene-2-carboxylic acid
CC1(C)[C@@H](O)CC[C@@]2(C)[C@H]1CC[C@]1(C)[C@@H]2C(=O)C=C2[C@@H]3C[C@](C)(CC[C@]3(C)CC[C@@]12C)C(O)=O
471-53-4
C30H46O4
470.69
MPDGHEJMBKOTSU-YKLVYJNSSA-N
(2S,4aS,6aS,6bR,8aR,10S,12aS,12bR,14bR)-10-hydroxy-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-icosahydropicene-2-carboxylic acid
CC1(C)[C@@H](O)CC[C@@]2(C)[C@H]1CC[C@]1(C)[C@@H]2C(=O)C=C2[C@@H]3C[C@](C)(CC[C@]3(C)CC[C@@]12C)C(O)=O

G2170, 471-53-4, C30H46O4

Spectrum™ 18beta-Glycyrrhetinic Acid , also known as Enoxolone, is often used to treat peptic ulcers and also has expectorant properties. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Packaging Amber Glass Bottle
Quantity 25 g
CAS Max % 1
Percent Purity Approx. 98%