Learn More
Justrite™ 19 Gallon ChemCor™ Under Fume Hood Corrosives/Acids Safety Cabinet With Manual-Closing Doors Available on GSA/VA Contract for Federal Government customers only.
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ 19 Gallon ChemCor™ Under Fume Hood Corrosives/Acids Safety Cabinet With Manual-Closing Doors provides superior corrosion protection as well as fire protection in a single cabinet.
$2940.00 - $3050.00
Specifications
Capacity 19 Gallon
Door Style Manual-close
View More Specs
Products 3
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
19-822-364 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Justrite™
8831022
Blue
Each for $2,940.00
Add to cart
19-822-365 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Justrite™
8831072
Light Neutral
Each for $3,030.00
Add to cart
19-822-366 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Justrite™
8831042
Silver
Each for $3,050.00
Add to cart
Description
Description
  • Expand storage, add new convenience to existing cabinets, or customize your new cabinet with additional shelves to meet specific workflow needs
  • Galvanized steel shelves hold up to 250 lbs and adjust on 3 in. increments for versatile storage
  • Patented SpillSlope galvanized steel shelves direct spills to back and bottom of a leakproof sump
  • Dispensing Shelf allows easy dispensing of liquids from a faucet without having to adjust the can's position on the shelf
  • The 11 in. depth allows the faucet to protrude beyond the shelf
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
19 Gallon
Manual-close