Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 19(R)-Hydroxyprostaglandin E2, 0.05% w/v in ethanol

Click to view available options
Quantity:
50μg
100μg
64625-54-3
C20H32O6
368.47
MFCD00135220
WTJYDBMHYPQFNJ-ZUVVJKHESA-N
19-hydroxy-pge2,19 r-hydroxy-prostaglandin e2,19 r-hydroxy-pge2,19r-hydroxy-pge2,19r-19-hydroxy pge-2,19-oh-pge2,19 r-oh-pge2,19r-oh-pge2,19 r-hydroxy prostaglandin e2,9-oxo-11r,15s,19r-trihydroxy-5z,13e-prostadienoic acid
5283038
(Z)-7-[(1R,2R,3R)-2-[(E,3S,7R)-3,7-dihydroxyoct-1-enyl]-3-hydroxy-5-oxocyclopentyl]hept-5-enoic acid
CC(CCCC(C=CC1C(CC(=O)C1CC=CCCCC(=O)O)O)O)O
64625-54-3
C20H32O6
368.47
MFCD00135220
WTJYDBMHYPQFNJ-ZUVVJKHESA-N
19-hydroxy-pge2,19 r-hydroxy-prostaglandin e2,19 r-hydroxy-pge2,19r-hydroxy-pge2,19r-19-hydroxy pge-2,19-oh-pge2,19 r-oh-pge2,19r-oh-pge2,19 r-hydroxy prostaglandin e2,9-oxo-11r,15s,19r-trihydroxy-5z,13e-prostadienoic acid
5283038
(Z)-7-[(1R,2R,3R)-2-[(E,3S,7R)-3,7-dihydroxyoct-1-enyl]-3-hydroxy-5-oxocyclopentyl]hept-5-enoic acid
CC(CCCC(C=CC1C(CC(=O)C1CC=CCCCC(=O)O)O)O)O
Quantity 100μg
UN Number UN1170
Concentration or Composition (by Analyte or Components) 0.05% w/v in ethanol
Formula Weight 370.49
Physical Form Liquid
Chemical Name or Material 19(R)-Hydroxyprostaglandin E2

GHS H Statement
H225-H371
Highly flammable liquid and vapor.
May cause damage to organs.

GHS P Statement
P210-P260-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: ETHANOL SOLUTIONS

storageNote1 : -30° to -10°C