Learn More
STERIS™ One Solution st™ Water For Injection
A diluent for surface sterilants, disinfectants, sanitizers, or cleaners used in cleanrooms or other critical environments. Meets USP specifications and is double bagged for easy entry into cleanrooms or aseptic areas.
Supplier: STERIS™ 1S03E4WR
Description
- Intended for diluting concentrated cleaners or disinfectants
- Filtered, and filled in a controlled environment
- Each package is double bagged and gamma irradiated for easy entry into controlled environments and aseptic core areas
- Includes an extensive documentation package to meet validation objectives
- Products meet United States Pharmacopeia (USP) Water For Injection (WFI) specifications
- Supported by the STERIS Technical Services team for individualized application and validation assistance
Specifications
Water for Injection (WFI) | |
Meets United States Pharmacopeia (USP) Water For Injection (WFI) Specifications | |
Diluting Concentrated Cleaners or Disinfectants | |
2/Pk. |
Sterile | |
Filtered | |
Double Bagged | |
2 gal. |