Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scott Safety™ AV-3000™ Facepiece with SureSeal

Catalog No. 19500631
Click to view available options
Includes:
Large dual voicemitters
Large dual voicemitters, communication bracket
Format:
Small
Medium
Large
Material:
Dupont™ Kevlar™ harness, Polyisoprene Seal
Polyester harness, Polyisoprene Seal
Rubber harness, Polyisoprene Seal
This item is not returnable. View return policy
This item is not returnable. View return policy

Requires

To use as an air-purifying respirator, requires cartridge adapter 805622-01 for 742 Series cartridges, 805059-01 for 40mm NIOSH cartridges and 200423-01 for CBRN Cap-1 canister.

Top-Down Convertibility design allows a single facepiece for APR, PAPR, SAR and SCBA respirators.

  • One fit test and reduced training
  • Patent-pending SureSeal technology for the most advanced fit and comfort
  • Wide seal offers a more secure interface with protective suits, hoods, shrouds and helmets
  • Mechanical and extensive electronic communication options across all applications
  • All components are latex free
  • Kevlar™, polyester or rubber head harness options with three nosecup sizes

Compliance:  
All head harnesses meet ANSI Z87.1-2003. Rubber and Kevlar head harnesses also meet NFPA 1981:2007. All head harnesses NIOSH approved for APR, supplied air, SCBA and PAPR respirators. Rubber and Kevlar head harnesses also NIOSH approved for CBRN respirators.

 

Order Info

Fit testing required prior to using this facepiece. Prescription lens kit (805753-01) and other accessories available separately.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance NIOSH, ANSI Z87.1-2003, NFPA 1981:2007
Material Dupont™ Kevlar™ harness, Polyisoprene Seal
Product Type Facepiece
Format Medium
For Use With (Equipment) Av-3000™ facepiece
Includes Large dual voicemitters
Latex Free Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.