Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H-1,2,4-Triazole-3-thiol 97%, ACROS Organics™

Click to view available options
Quantity:
10g
100g
Packaging:
Glass bottle
3179-31-5
C2H3N3S
101.127
MFCD00005229
AFBBKYQYNPNMAT-UHFFFAOYSA-N
1h-1,2,4-triazole-3-thiol,3-mercapto-1,2,4-triazole,1,2,4-triazole-3-thiol,mercaptotriazole,4h-1,2,4-triazole-3-thiol,s-triazole-3-thiol,3-mercapto-1h-1,2,4-triazole,1,2,4-triazole-5-thiol,1,3,4-triazine-2-thiol,1,3,4-triazole-2-thiol
2723802
1,2-dihydro-1,2,4-triazole-3-thione
C1=NC(=S)NN1
3179-31-5
C2H3N3S
101.127
MFCD00005229
AFBBKYQYNPNMAT-UHFFFAOYSA-N
1h-1,2,4-triazole-3-thiol,3-mercapto-1,2,4-triazole,1,2,4-triazole-3-thiol,mercaptotriazole,4h-1,2,4-triazole-3-thiol,s-triazole-3-thiol,3-mercapto-1h-1,2,4-triazole,1,2,4-triazole-5-thiol,1,3,4-triazine-2-thiol,1,3,4-triazole-2-thiol
2723802
1,2-dihydro-1,2,4-triazole-3-thione
C1=NC(=S)NN1
Infrared Spectrum Authentic
Melting Point 217°C to 220°C
Color White to Yellow
Quantity 10g
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 26, 142
Formula Weight 101.13
Physical Form Fine Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 1H-1, 2, 4-Triazole-3-thiol, 97%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

STENCH

EINECSNumber : 221-656-6

RTECSNumber : XZ5267500

TSCA : TSCA