Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H,1H,2H,2H-Heptadecafluorodecyl Acrylate (stabilized with BHT + TBC) 97.0+%, TCI America™

Click to view available options
Quantity:
10g
27905-45-9
C13H7F17O2
518.17
MFCD00042306
QUKRIOLKOHUUBM-UHFFFAOYSA-N
1h,1h,2h,2h-heptadecafluorodecyl acrylate,1h,1h,2h,2h-perfluorodecyl acrylate,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl acrylate,unii-y9bfb4689z,2-perfluorooctyl ethyl acrylate,1,1,2,2-tetrahydroperfluorodecyl acrylate,perfluorooctyl ethyl acrylate,2-propenoic acid, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl ester,2-perfluorooctyl ethylacrylate,c8f17ch2ch2ococh=ch2
119747
3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl prop-2-enoate
C=CC(=O)OCCC(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
27905-45-9
C13H7F17O2
518.17
MFCD00042306
QUKRIOLKOHUUBM-UHFFFAOYSA-N
1h,1h,2h,2h-heptadecafluorodecyl acrylate,1h,1h,2h,2h-perfluorodecyl acrylate,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl acrylate,unii-y9bfb4689z,2-perfluorooctyl ethyl acrylate,1,1,2,2-tetrahydroperfluorodecyl acrylate,perfluorooctyl ethyl acrylate,2-propenoic acid, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl ester,2-perfluorooctyl ethylacrylate,c8f17ch2ch2ococh=ch2
119747
3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl prop-2-enoate
C=CC(=O)OCCC(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
Boiling Point 90°C
Color Colorless
Quantity 10g
Formula Weight 518.17
Physical Form Clear Liquid at 20°C
Percent Purity ≥97.0% (GC)
Chemical Name or Material 1H,1H,2H,2H-Heptadecafluorodecyl Acrylate (stabilized with BHT + TBC)

EINECSNumber : (2)-3502

TSCA : Yes