Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H,1H,2H,2H-Perfluoro-1-iodohexane, 97%, Thermo Scientific Chemicals

Catalog Number p-7027372
Click to view available options
Quantity:
5 g
25 g
2043-55-2
C6H4F9I
373.988
MFCD00039409
CXHFIVFPHDGZIS-UHFFFAOYSA-N
1h,1h,2h,2h-perfluorohexyl iodide, 2-perfluorobutyl ethyl iodide, hexane, 1,1,1,2,2,3,3,4,4-nonafluoro-6-iodo, 1h,1h,2h,2h-nonafluorohexyl iodide, 1-iodo-1h,1h,2h,2h-nonafluorohexane, 1,1,2,2-tetrahydroperfluorohexyliodide, 2-nonafluorobutyl ethyl iodide, 1-iodo-1h,1h,2h,2h-perfluorohexane, 1h,1h,2h,2h-perfluoro-1-iodohexane, 1,1,1,2,2,3,3,4,4-nonafluoro-6-iodo-hexane
74887
1,1,1,2,2,3,3,4,4-nonafluoro-6-iodohexane
C(CI)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F
2043-55-2
C6H4F9I
373.988
MFCD00039409
CXHFIVFPHDGZIS-UHFFFAOYSA-N
1h,1h,2h,2h-perfluorohexyl iodide, 2-perfluorobutyl ethyl iodide, hexane, 1,1,1,2,2,3,3,4,4-nonafluoro-6-iodo, 1h,1h,2h,2h-nonafluorohexyl iodide, 1-iodo-1h,1h,2h,2h-nonafluorohexane, 1,1,2,2-tetrahydroperfluorohexyliodide, 2-nonafluorobutyl ethyl iodide, 1-iodo-1h,1h,2h,2h-perfluorohexane, 1h,1h,2h,2h-perfluoro-1-iodohexane, 1,1,1,2,2,3,3,4,4-nonafluoro-6-iodo-hexane
74887
1,1,1,2,2,3,3,4,4-nonafluoro-6-iodohexane
C(CI)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -25°C
Density 1.94
Boiling Point 138°C to 140°C
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.37
Quantity 5 g
Beilstein 1870862
Sensitivity Light sensitive
Formula Weight 373.99
Percent Purity 97%
Chemical Name or Material 1H,1H,2H,2H-Perfluoro-1-iodohexane
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 218-055-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit