Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1H,1H,2H,2H-Perfluorodecyltriethoxysilane, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
25 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
1H,1H,2H,2H-Perfluorodecyltriethoxysilane is used to bond poly(tetrafluoroethylene) films to silicon wafers. It is also utilized in anti-reflective coating, release coating and soil repellent coating.
Solubility
Miscible with ethanol and tetrahydrofuran.
Notes
Incompatible with strong oxidizing agents.
1H,1H,2H,2H-Perfluorodecyltriethoxysilane is used to bond poly(tetrafluoroethylene) films to silicon wafers. It is also utilized in anti-reflective coating, release coating and soil repellent coating.
Solubility
Miscible with ethanol and tetrahydrofuran.
Notes
Incompatible with strong oxidizing agents.
Specifications
Boiling Point | 103°C to 106°C (3mmHg) |
Quantity | 1 g |
Linear Formula | CF3(CF2)7(CH2)2Si(OCH2CH3)3 |
UN Number | UN1760 |
Flash Point | 100°C (212°F) |
Solubility Information | Miscible with ethanol and tetrahydrofuran. |
Formula Weight | 610.38 |
Percent Purity | 97% |
Refractive Index | 1.342 |
Sensitivity | Moisture sensitive |
Show More |
Safety and Handling
Hazard Category | H314-H335 |
Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S. |
TSCA | No |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction