Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1H,1H,2H,2H-Perfluorooctyltriethoxysilane, 97%

Click to view available options
Quantity:
5g
25g
51851-37-7
C14H19F13O3Si
510.367
MFCD00042333
AVYKQOAMZCAHRG-UHFFFAOYSA-N
1h,1h,2h,2h-perfluorooctyltriethoxysilane,triethoxy 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane,silane, triethoxy 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl,perfluorooctyl triethoxysilane,3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyltriethoxysilan,triethoxy 1h,1h,2h,2h-perfluoro-1-octyl silane,tridecafluoro-1,1,2,2-tetrahydrooctyl triethoxysilane,1h,1h,2h,2h-perfluoroalkyltriethoxysilane,triethoxy-1h,1h,2h,2h-perfluoro-n-octylsilane
103991
triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane
CCO[Si](CCC(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(OCC)OCC
51851-37-7
C14H19F13O3Si
510.367
MFCD00042333
AVYKQOAMZCAHRG-UHFFFAOYSA-N
1h,1h,2h,2h-perfluorooctyltriethoxysilane,triethoxy 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane,silane, triethoxy 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl,perfluorooctyl triethoxysilane,3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyltriethoxysilan,triethoxy 1h,1h,2h,2h-perfluoro-1-octyl silane,tridecafluoro-1,1,2,2-tetrahydrooctyl triethoxysilane,1h,1h,2h,2h-perfluoroalkyltriethoxysilane,triethoxy-1h,1h,2h,2h-perfluoro-n-octylsilane
103991
triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane
CCO[Si](CCC(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(OCC)OCC
Applications
1H,1H,2H,2H-Perfluorooctyltriethoxysilane is used as an active component of fluorine modified ORMOCER (organically modified ceramics) coating materials. It is a fluorine based polymer having low surface free energy, enhances its anti-adhesive behavior of polar and non-polar substances. Its wetting property acts in repelling aqueous electrolyte solution away from metallic substrates, thereby giving protection to metal substrate from corrosion. It is utilized in the surface modification of glass nanoparticles.

Solubility
Miscible with alcohol, aromatic and aliphatic hydrocarbons.

Notes
Moisture sensitive. Incompatible with moisture and oxidizing agents.
Linear Formula CF3(CF2)5(CH2)2Si(OCH2CH3)3
Formula Weight 510.37
Percent Purity 97%
Boiling Point 220°C
Melting Point <-38°C
Quantity 25g
Flash Point 97°C (206°F)
Chemical Name or Material 1H,1H,2H,2H-Perfluorooctyltriethoxysilane

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

TSCA : TSCA