Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H,1H-Tridecafluoro-1-heptanol 95.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
375-82-6
C7H3F13O
350.079
MFCD00042433
STLNAVFVCIRZLL-UHFFFAOYSA-N
1H,1H-Perfluoro-1-heptanol
550386
2,2,3,3,4,4,5,5,6,6,7,7,7-tridecafluoroheptan-1-ol
C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O
375-82-6
C7H3F13O
350.079
MFCD00042433
STLNAVFVCIRZLL-UHFFFAOYSA-N
1H,1H-Perfluoro-1-heptanol
550386
2,2,3,3,4,4,5,5,6,6,7,7,7-tridecafluoroheptan-1-ol
C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O
Boiling Point 95°C
Color Yellow
Quantity 25g
Formula Weight 350.08
Physical Form Clear Liquid at 20°C
Percent Purity ≥95.0% (GC)
Chemical Name or Material 1H,1H-Tridecafluoro-1-heptanol