Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H-Benzimidazole-2-ethanamine dihydrochloride, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
4500-12-3
C10H15NO
165.24
MFCD01325081
RABVIFXMFZFITE-KHPPLWFESA-N
2-adamantanone oxime,adamantan-2-one oxime,adamantanone oxime,2-adamantanone, oxime,2-hydroxyiminoadamantane,adamantanone-2-oxime,2-isonitrosoadamantane,2-hydroxyimino adamantane,n-2-adamantylidene hydroxylamine,n-adamantan-2-ylidene hydroxylamine
64158
N-(2-adamantylidene)hydroxylamine
C1C2CC3CC1CC(C2)C3=NO
4500-12-3
C10H15NO
165.24
MFCD01325081
RABVIFXMFZFITE-KHPPLWFESA-N
2-adamantanone oxime,adamantan-2-one oxime,adamantanone oxime,2-adamantanone, oxime,2-hydroxyiminoadamantane,adamantanone-2-oxime,2-isonitrosoadamantane,2-hydroxyimino adamantane,n-2-adamantylidene hydroxylamine,n-adamantan-2-ylidene hydroxylamine
64158
N-(2-adamantylidene)hydroxylamine
C1C2CC3CC1CC(C2)C3=NO
Packaging Glass bottle
Color White to Gray
Infrared Spectrum Authentic
Quantity 1g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 234.13
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 1H-Benzimidazole-2-ethanamine dihydrochloride, 97%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.