Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1H-Benzotriazol-1-yloxytri(1-pyrrolidinyl)phosphonium hexafluorophosphate, 98%

Click to view available options
Quantity:
5g
25g
128625-52-5
C18H28F6N6OP2
520.401
MFCD00077411
VIAFLMPQBHAMLI-UHFFFAOYSA-N
pybop,1h-benzo d 1,2,3 triazol-1-yl oxy tri pyrrolidin-1-yl phosphonium hexafluorophosphate v,benzotriazol-1-yloxy tripyrrolidinophosphonium hexafluorophosphate,benzotriazole-1-yl-oxytripyrrolidinophosphonium hexafluorophosphate,benzotriazol-1-yl-oxytripyrrolidinophosphonium hexafluorophosphate,1h-benzotriazol-1-yloxytripyrrolidinophosphonium hexafluorophosphate,benzotriazol-1-yloxytris pyrrolidino phosphonium hexafluorophosphate,benzotriazole-1-yl-oxy-tris-pyrrolidino-phosphonium hexafluorophosphate,benzotriazol-1-yl-oxy-tripyrrolidinophosphonium hexafluorophosphate,benzotriazol-1-yl-oxytripyrrolidinphosphonium hexafluorophosphate
2724699
benzotriazol-1-yloxy(tripyrrolidin-1-yl)phosphanium;hexafluorophosphate
C1CCN(C1)[P+](N2CCCC2)(N3CCCC3)ON4C5=CC=CC=C5N=N4.F[P-](F)(F)(F)(F)F
128625-52-5
C18H28F6N6OP2
520.401
MFCD00077411
VIAFLMPQBHAMLI-UHFFFAOYSA-N
pybop,1h-benzo d 1,2,3 triazol-1-yl oxy tri pyrrolidin-1-yl phosphonium hexafluorophosphate v,benzotriazol-1-yloxy tripyrrolidinophosphonium hexafluorophosphate,benzotriazole-1-yl-oxytripyrrolidinophosphonium hexafluorophosphate,benzotriazol-1-yl-oxytripyrrolidinophosphonium hexafluorophosphate,1h-benzotriazol-1-yloxytripyrrolidinophosphonium hexafluorophosphate,benzotriazol-1-yloxytris pyrrolidino phosphonium hexafluorophosphate,benzotriazole-1-yl-oxy-tris-pyrrolidino-phosphonium hexafluorophosphate,benzotriazol-1-yl-oxy-tripyrrolidinophosphonium hexafluorophosphate,benzotriazol-1-yl-oxytripyrrolidinphosphonium hexafluorophosphate
2724699
benzotriazol-1-yloxy(tripyrrolidin-1-yl)phosphanium;hexafluorophosphate
C1CCN(C1)[P+](N2CCCC2)(N3CCCC3)ON4C5=CC=CC=C5N=N4.F[P-](F)(F)(F)(F)F
Applications
1H-Benzotriazol-1-yloxytri(1-pyrrolidinyl) phosphonium hexafluorophosphate is a peptide coupling reagent utilized in solid phase peptide synthesis. It is also used as a substitute for the (benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP) reagent.

Solubility
Soluble in chloroform.

Notes
Store in a cool place. Incompatible with oxidizing agents and water.
Quantity 25g
Melting Point ~155°C (decomposition)
Beilstein 3584612
Formula Weight 520.4
Percent Purity 98%
Chemical Name or Material 1H-Benzotriazol-1-yloxytri(1-pyrrolidinyl)phosphonium hexafluorophosphate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C