Alfa Aesar™ 1H-Benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate, 98%

Manufacturer: Alfa Aesar™ A1614003

Quantity 1g

 View more versions of this product

Catalog No. AAA1614003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 56602-33-6
    Molecular Formula C12H22F6N6OP2
    Formula Weight 442.29
    MDL Number MFCD00011948
    UN Number UN1325
    Beilstein 4287025
    Synonym BOP Reagent; Castro's Reagent
    Chemical Name or Material 1H-Benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate

    Specifications

    Formula Weight 442.29
    MDL Number MFCD00011948
    UN Number UN1325
    Synonym BOP Reagent; Castro's Reagent
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H228-H315-H319-H335
    Flammable solid.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Recommended Storage 2° to 8°C
    Melting Point ≈138°C (dec.)
    Quantity 1g
    Molecular Formula C12H22F6N6OP2
    CAS 56602-33-6
    Beilstein 4287025
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
    Chemical Name or Material 1H-Benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate
    Assay 98%

    Description

    Applications
    1H-Benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate is used as a reagent for peptide coupling, lactonization, selective esterification, amidation of alfa amino acids without racemization and synthesis of magnolamide for antioxidative activity and catalyst for 9-acridinecaroboxamide derivative. It is also used as a precursor for the synthesis of phenyl esters of amino acids. It acts as a substitute for (Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP) reagent.

    Solubility
    Soluble in methanol, acetone and dichloromethane. Insoluble in water.

    Notes
    Moisture and light sensitive. Incompatible with strong oxidizing agents. Store in a cool place. Keep the container tightly closed.

    GHS H Statement
    H228-H315-H319-H335
    Flammable solid.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2° to 8°C

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit