Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H-Indazol-6-ylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
885068-10-0
C7H7BN2O2
161.96
MFCD04972419
ZKNLCHWRWRYPGG-UHFFFAOYSA-N
6-indazolyboronic acid,1h-indazol-6-yl-6-boronic acid,1h-indazole-6-boronic acid,indazole-6-boronic acid,1h-indazol-6-yl boronic acid,6-indazolboronic acid,boronic acid, 1h-indazol-6-yl,1h-indazole-6-boronicacid,6-indazoleboronicacid,zlchem 1076
24728617
1H-indazol-6-ylboronic acid
B(C1=CC2=C(C=C1)C=NN2)(O)O
885068-10-0
C7H7BN2O2
161.96
MFCD04972419
ZKNLCHWRWRYPGG-UHFFFAOYSA-N
6-indazolyboronic acid,1h-indazol-6-yl-6-boronic acid,1h-indazole-6-boronic acid,indazole-6-boronic acid,1h-indazol-6-yl boronic acid,6-indazolboronic acid,boronic acid, 1h-indazol-6-yl,1h-indazole-6-boronicacid,6-indazoleboronicacid,zlchem 1076
24728617
1H-indazol-6-ylboronic acid
B(C1=CC2=C(C=C1)C=NN2)(O)O
Infrared Spectrum Authentic
Melting Point 180.0°C to 184.0°C
Quantity 5g
Color White
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 161.96
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 1H-Indazol-6-ylboronic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning