Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H-Indazole-4-carboxaldehyde, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
669050-70-8
C8H6N2O
146.15
MFCD06738279
FPJXNCKSPFGQGC-UHFFFAOYSA-N
4-formylindazole,1h-indazole-4-carboxaldehyde,4-formyl-1h-indazole,1h-indazol-4-carboxaldehyde,pubchem22512,acmc-209nwx,1h-indazole-4-carboxaldehyde 9ci
45083569
1H-indazole-4-carbaldehyde
C1=CC(=C2C=NNC2=C1)C=O
669050-70-8
C8H6N2O
146.15
MFCD06738279
FPJXNCKSPFGQGC-UHFFFAOYSA-N
4-formylindazole,1h-indazole-4-carboxaldehyde,4-formyl-1h-indazole,1h-indazol-4-carboxaldehyde,pubchem22512,acmc-209nwx,1h-indazole-4-carboxaldehyde 9ci
45083569
1H-indazole-4-carbaldehyde
C1=CC(=C2C=NNC2=C1)C=O
Color White
Melting Point 166.0°C to 167.0°C
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 146.15
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 1H-Indazole-4-carboxaldehyde
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning