Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H-Indazole, 99%, Thermo Scientific Chemicals

Catalog Number p-7081619
Click to view available options
Quantity:
1 g
5 g
25 g
271-44-3
C7H6N2
118.139
MFCD00005691
BAXOFTOLAUCFNW-UHFFFAOYSA-N
indazole, 1h-benzopyrazole, 2-azaindole, isoindazole, 2h-indazole, 1,2-diazaindene, 1,2-benzodiazole, 1h-indazol, benzopyrazole, 1,2-benzopyrazole
9221
CHEBI:36669
1H-indazole
C1=CC=C2C(=C1)C=NN2
271-44-3
C7H6N2
118.139
MFCD00005691
BAXOFTOLAUCFNW-UHFFFAOYSA-N
indazole, 1h-benzopyrazole, 2-azaindole, isoindazole, 2h-indazole, 1,2-diazaindene, 1,2-benzodiazole, 1h-indazol, benzopyrazole, 1,2-benzopyrazole
9221
CHEBI:36669
1H-indazole
C1=CC=C2C(=C1)C=NN2

1H-Indazole is used in organic synthesis, it can react with butyryl chloride and 1-butyryl-1H-indazole. It is also used in synthesis of several small molecule inhibitors as potential cancer therapeutics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H-Indazole is used in organic synthesis, it can react with butyryl chloride and 1-butyryl-1H-indazole. It is also used in synthesis of several small molecule inhibitors as potential cancer therapeutics.

Solubility
Soluble in hot water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Melting Point 146°C to 149°C
Boiling Point 270°C
Quantity 1 g
Beilstein 109676
Merck Index 14,4936
Solubility Information Soluble in hot water.
Formula Weight 118.14
Percent Purity 99%
Chemical Name or Material 1H-Indazole
EINECSNumber 205-978-4
RTECSNumber NK7745000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit