Learn More
Medchemexpress LLC 1H-Indole-3-carboxylic acid, methyl ester | 942-24-5 | MFCD00189407 | 99.9% | C10H9NO2 | 50 G
SDP
Supplier:  Medchemexpress LLC HY7963550G
Encompass_Preferred
Methyl indole-3-carboxylate is a Sorangium metabolite that shows weak activity against Gram-positive Nocardia sp. with a MIC of 33.3 μg/mL. This product is for research use only and is not sold to patients. It can be dissolved in DMSO at 100 mg/mL for in vitro use and various protocols are available for in vivo dissolution based on experimental animal and administration route.
  • Sorangium metabolite
  • Weak activity against Gram-positive Nocardia sp. (MIC of 33.3 μg/mL)
  • No significant cytotoxicity against L929, PC-3, and KB3-1 cells (IC50 > 56.8 μM)
  • Soluble in DMSO at 100 mg/mL for in vitro experiments
  • Multiple in vivo dissolution protocols available
  • Store as powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-32723