Alfa Aesar™ 1H-Tetrazole-5-carboxylic acid ethyl ester sodium salt, 97%

Manufacturer: Alfa Aesar™ B2346122

Quantity 100g

 View more versions of this product

Catalog No. AAB2346122

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 96107-94-7
    Molecular Formula C4H5N4NaO2
    Formula Weight 164.11
    MDL Number MFCD02683105
    UN Number UN1325
    Chemical Name or Material 1H-Tetrazole-5-carboxylic acid ethyl ester sodium salt

    Specifications

    Formula Weight 164.11
    Assay Percent Range 0.99
    Melting Point ≈240°C (dec.)
    Molecular Formula C4H5N4NaO2
    MDL Number MFCD02683105
    CAS 96107-94-7
    UN Number UN1325
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H228
    Flammable solid.
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P241-P370+P378a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    Use explosion-proof electrical/ventilating/lighting/ equipment.
    In case of fire: Use for extinction: CO2, powder or water spray.
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
    Quantity 100g
    Chemical Name or Material 1H-Tetrazole-5-carboxylic acid ethyl ester sodium salt
    Assay 97%

    GHS H Statement
    H228
    Flammable solid.

    GHS P Statement
    P210-P280-P240-P241-P370+P378a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    Use explosion-proof electrical/ventilating/lighting/ equipment.
    In case of fire: Use for extinction: CO2, powder or water spray.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit