Learn More
Leica Biosystems 1ml NCL-DYSB
Supplier:  Leica Biosystems DYSB
Encompass
Antigen Background Duchenne Muscular dystrophy (DMD) is the most common of the muscular dystrophies resulting in progressive muscular wasting and death. Dystrophin is the 427kD protein product of the DMD gene located on the X chromosome at position Xp21. Abnormalities in protein expression occur in patients with DMD/BMD and dystrophin analysis may be used to distinguish these conditions from other neuromuscular diseases. Severe Duchenne muscular dystrophy is associated with a marked dystrophin deficiency, whereas patients with the milder form of Becker muscular dystrophy show less pronounced abnormalities of protein expression. The immunolabeling patterns for DYS1, DYS2 and DYS3 are similar; however, the use of all three antibodies is recommended to avoid the possibility of occasional false negative results. Disclaimer Dystrophin Antibodies are recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology
Catalog No. 50-255-2265
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.