Learn More
Abnova™ 1p36.3 (Orange) FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Manufacturer:  Abnova™ FG0103
Catalog No. 89-932-016
Add to cart
Description
Description
Specifications
Specifications
FISH
200 µL
Store at -20°C in the dark.