Alfa Aesar™ (1R)-10-Camphorsulfonamide, 97%

Manufacturer: Alfa Aesar™ 4435814

Quantity 25g

 View more versions of this product

Catalog No. AA4435814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 72597-34-3
    Molecular Formula C10H17NO3S
    Formula Weight 231.32
    MDL Number MFCD00151500
    Chemical Name or Material (1R)-10-Camphorsulfonamide

    Specifications

    Formula Weight 231.32
    Melting Point 134°C (dec.)
    CAS 72597-34-3
    MDL Number MFCD00151500
    Quantity 25g
    Physical Form Powder
    Molecular Formula C10H17NO3S
    Chemical Name or Material (1R)-10-Camphorsulfonamide
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit