Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,2R)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol, 96%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
716-61-0
C9H12N2O4
212.2
MFCD00078126
OCYJXSUPZMNXEN-RKDXNWHRSA-N
1r,2r-2-amino-1-4-nitrophenyl propane-1,3-diol,chloramphenicol base,levoamine,1r,2r---2-amino-1-4-nitrophenyl-1,3-propanediol,d---threo-2-amino-1-4-nitrophenyl-1,3-propanediol,d-threo---2-amino-1-p-nitrophenyl-1,3-propanediol,1r,2r-2-amino-1-4-nitrophenyl-1,3-propanediol,d--threo-2-amino-1-p-nitrophenyl propane-1,3-diol,1,3-propanediol, 2-amino-1-4-nitrophenyl-, 1r,2r,1,3-propanediol, 2-amino-1-p-nitrophenyl-, d-threo--
69745
CHEBI:78660
C1=CC(=CC=C1C(C(CO)N)O)[N+](=O)[O-]
716-61-0
C9H12N2O4
212.2
MFCD00078126
OCYJXSUPZMNXEN-RKDXNWHRSA-N
1r,2r-2-amino-1-4-nitrophenyl propane-1,3-diol,chloramphenicol base,levoamine,1r,2r---2-amino-1-4-nitrophenyl-1,3-propanediol,d---threo-2-amino-1-4-nitrophenyl-1,3-propanediol,d-threo---2-amino-1-p-nitrophenyl-1,3-propanediol,1r,2r-2-amino-1-4-nitrophenyl-1,3-propanediol,d--threo-2-amino-1-p-nitrophenyl propane-1,3-diol,1,3-propanediol, 2-amino-1-4-nitrophenyl-, 1r,2r,1,3-propanediol, 2-amino-1-p-nitrophenyl-, d-threo--
69745
CHEBI:78660
C1=CC(=CC=C1C(C(CO)N)O)[N+](=O)[O-]
Specific Rotation -31
Color Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25g
Melting Point 160°C to 166°C
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Formula Weight 212.2
Physical Form Powder
Percent Purity ≥95%
Chemical Name or Material (1R, 2R)-2-Amino-1-(4-nitrophenyl)propane-1, 3-diol, 96%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 211-938-7

RTECSNumber : TY3100000

TSCA : TSCA