Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,2R,3R,5S)-(-)-Isopinocampheylamine, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
69460-11-3
C10H19N
153.27
VPTSZLVPZCTAHZ-BZNPZCIMSA-N
1r,2r,3r,5s---isopinocampheylamine,1r,2r,3r,5s-2,6,6-trimethylbicyclo 3.1.1 heptan-3-amine,bicyclo 3.1.1 heptan-3-amine, 2,6,6-trimethyl-, 1r,2r,3r,5s,--isopinocampheylamine,--isopinocampheyl amine,1r,2r,3r,5s-3-pinanamine,1r,2r,5s-pinane-3alpha-amine,--isopinocampheylamine; 1r,2r,3r,5s-3-pinanamine,1r,2r,3r,5s-2,6,6-trimeth-ylbicyclo 3.1.1 heptane-3-amine
11744832
(1S,3R,4R,5R)-4,6,6-trimethylbicyclo[3.1.1]heptan-3-amine
CC1C2CC(C2(C)C)CC1N
69460-11-3
C10H19N
153.27
VPTSZLVPZCTAHZ-BZNPZCIMSA-N
1r,2r,3r,5s---isopinocampheylamine,1r,2r,3r,5s-2,6,6-trimethylbicyclo 3.1.1 heptan-3-amine,bicyclo 3.1.1 heptan-3-amine, 2,6,6-trimethyl-, 1r,2r,3r,5s,--isopinocampheylamine,--isopinocampheyl amine,1r,2r,3r,5s-3-pinanamine,1r,2r,5s-pinane-3alpha-amine,--isopinocampheylamine; 1r,2r,3r,5s-3-pinanamine,1r,2r,3r,5s-2,6,6-trimeth-ylbicyclo 3.1.1 heptane-3-amine
11744832
(1S,3R,4R,5R)-4,6,6-trimethylbicyclo[3.1.1]heptan-3-amine
CC1C2CC(C2(C)C)CC1N
Specific Rotation -42
Flash Point 72°C
Boiling Point 90°C (18.0mmHg)
Color Colorless to Yellow
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Formula Weight 153.27
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material (1R,2R,3R,5S)-(-)-Isopinocampheylamine
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning