Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,2R)-Bis[(2-methoxyphenyl)phenylphosphino]ethane, 90%, Acros Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
55739-58-7
C28H28O2P2
458.48
MFCD05863546
QKZWXPLBVCKXNQ-ROJLCIKYSA-N
r,r-dipamp,1,2-bis r-2-methoxyphenyl phenyl phosphino ethane,unii-blj831owlw,--dipamp,r,r-ethylenebis 2-methoxyphenyl phenylphosphine,r,r-1,2-bis 2-methoxyphenyl phenylphosphino ethane,blj831owlw,s,s-dipamp,1r,2r---bis 2-methoxyphenyl phenylphosphino ethane
10884975
(R)-(2-methoxyphenyl)-[2-[(2-methoxyphenyl)-phenylphosphanyl]ethyl]-phenylphosphane
COC1=CC=CC=C1P(CCP(C2=CC=CC=C2)C3=CC=CC=C3OC)C4=CC=CC=C4
55739-58-7
C28H28O2P2
458.48
MFCD05863546
QKZWXPLBVCKXNQ-ROJLCIKYSA-N
r,r-dipamp,1,2-bis r-2-methoxyphenyl phenyl phosphino ethane,unii-blj831owlw,--dipamp,r,r-ethylenebis 2-methoxyphenyl phenylphosphine,r,r-1,2-bis 2-methoxyphenyl phenylphosphino ethane,blj831owlw,s,s-dipamp,1r,2r---bis 2-methoxyphenyl phenylphosphino ethane
10884975
(R)-(2-methoxyphenyl)-[2-[(2-methoxyphenyl)-phenylphosphanyl]ethyl]-phenylphosphane
COC1=CC=CC=C1P(CCP(C2=CC=CC=C2)C3=CC=CC=C3OC)C4=CC=CC=C4
Specific Rotation -80° to -85° (20°C, 589nm) (C=1, CHCl3)
Packaging Glass bottle
Quantity 250mg
Infrared Spectrum Authentic
Color White
Melting Point 102.0°C to 104.0°C
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 90%
Merck Index 15, 3333
Formula Weight 458.48
Physical Form Crystalline Powder
Percent Purity 90%
Chemical Name or Material (1R,2R)-Bis[(2-methoxyphenyl)phenylphosphino]ethane
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning