Alfa Aesar™ (1R,2R)-N-Methylsulfonyl-1,2-diphenylethanediamine, 98+%

Manufacturer: Alfa Aesar™ H2770406

Quantity 5g

 View more versions of this product

Catalog No. AAH2770406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 511534-44-4
    Molecular Formula C15H18N2O2S
    Formula Weight 290.38
    MDL Number MFCD03791036
    UN Number UN3259
    Synonym (1R,2R)-N-Methanesulfonyl-1,2-diphenylethanediamine
    Chemical Name or Material (1R,2R)-N-Methylsulfonyl-1,2-diphenylethanediamine

    Specifications

    Formula Weight 290.38
    MDL Number MFCD03791036
    UN Number UN3259
    Melting Point 117° to 119°C
    Quantity 5g
    Molecular Formula C15H18N2O2S
    CAS 511534-44-4
    Synonym (1R,2R)-N-Methanesulfonyl-1,2-diphenylethanediamine
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
    Chemical Name or Material (1R,2R)-N-Methylsulfonyl-1,2-diphenylethanediamine
    Assay ≥98%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit