Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,2R)-N,N-Bis(2-(diphenylphosphino)benzyl)cyclohexane-1,2-diamine, 95%, ACROS Organics™

Click to view available options
Quantity:
250mg
174758-63-5
C44H44N2P2
662.79
MFCD16618374
OBHPYVNBXWUKNY-NCRNUEESSA-N
1r,2r-n1,n2-bis 2-diphenylphosphino benzyl cyclohexane-1,2-diamine,1r,2r-n,n-bis 2-diphenylphosphino benzyl cyclohexane-1,2-diamine,1r,2r-n,n'-bis 2-diphenylphosphino benzyl cyclohexane-1,2-diamine,1r,2r-n1,n2-bis 2-diphenylphosphanyl phenyl methyl cyclohexane-1,2-diamine,1r,2r-1,2-bis 2-diphenylphosphino benzylamino cyclohexane,1r,2r-1-n,2-n-bis 2-diphenylphosphanylphenyl methyl cyclo,trans-n,n'-bis 2-diphenylphosphanylbenzyl cyclohexane-1,2-diamine,1r,2r-n1,n2-bis 2-diphenylphosphanyl benzyl cyclohexane-1,2-diamine
10909155
(1R,2R)-1-N,2-N-bis[(2-diphenylphosphanylphenyl)methyl]cyclohexane-1,2-diamine
C1CCC(C(C1)NCC2=CC=CC=C2P(C3=CC=CC=C3)C4=CC=CC=C4)NCC5=CC=CC=C5P(C6=CC=CC=C6)C7=CC=CC=C7
174758-63-5
C44H44N2P2
662.79
MFCD16618374
OBHPYVNBXWUKNY-NCRNUEESSA-N
1r,2r-n1,n2-bis 2-diphenylphosphino benzyl cyclohexane-1,2-diamine,1r,2r-n,n-bis 2-diphenylphosphino benzyl cyclohexane-1,2-diamine,1r,2r-n,n'-bis 2-diphenylphosphino benzyl cyclohexane-1,2-diamine,1r,2r-n1,n2-bis 2-diphenylphosphanyl phenyl methyl cyclohexane-1,2-diamine,1r,2r-1,2-bis 2-diphenylphosphino benzylamino cyclohexane,1r,2r-1-n,2-n-bis 2-diphenylphosphanylphenyl methyl cyclo,trans-n,n'-bis 2-diphenylphosphanylbenzyl cyclohexane-1,2-diamine,1r,2r-n1,n2-bis 2-diphenylphosphanyl benzyl cyclohexane-1,2-diamine
10909155
(1R,2R)-1-N,2-N-bis[(2-diphenylphosphanylphenyl)methyl]cyclohexane-1,2-diamine
C1CCC(C(C1)NCC2=CC=CC=C2P(C3=CC=CC=C3)C4=CC=CC=C4)NCC5=CC=CC=C5P(C6=CC=CC=C6)C7=CC=CC=C7
Melting Point >300.0°C
Color Yellow
Quantity 250mg
NMR (1H-NMR) Consistent with structure, (31P-NMR) Consistent with structure
Infrared Spectrum Authentic
Elemental Analysis 74.96 to 84.52% C, 3.98 to 4.48% N
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 662.79
Physical Form Crystalline Powder
Percent Purity 95%
Chemical Name or Material (1R,2R)-N,N-Bis(2-(Diphenylphosphino)benzyl)cyclohexane-1,2-diamine
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning