Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,2R)-N,N'-Dimethyl-1,2-cyclohexanediamine, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
68737-65-5
C8H18N2
142.24
MFCD00671527
JRHPOFJADXHYBR-HTQZYQBOSA-N
1r,2r-n,n'-dimethyl-1,2-cyclohexanediamine,1r,2r-n1,n2-dimethylcyclohexane-1,2-diamine,1r,2r---n,n'-dimethylcyclohexane-1,2-diamine,trans-n1,n2-dimethylcyclohexane-1,2-diamine,trans-1r,2r-n,n'-bismethyl-1,2-cyclohexanediamine,trans-n,n'-dimethylcyclohexane-1,2-diamine,trans-n,n'-dimethyl-1,2-cyclohexanediamine,trans-1,2-bis methylamino cyclohexane,trans-1r,2r-n,n'-bismethyl-1,2-cyclohexane diamine,1r,2r---1,2-bis methylamino cyclohexane
2733821
(1R,2R)-1-N,2-N-dimethylcyclohexane-1,2-diamine
CNC1CCCCC1NC
68737-65-5
C8H18N2
142.24
MFCD00671527
JRHPOFJADXHYBR-HTQZYQBOSA-N
1r,2r-n,n'-dimethyl-1,2-cyclohexanediamine,1r,2r-n1,n2-dimethylcyclohexane-1,2-diamine,1r,2r---n,n'-dimethylcyclohexane-1,2-diamine,trans-n1,n2-dimethylcyclohexane-1,2-diamine,trans-1r,2r-n,n'-bismethyl-1,2-cyclohexanediamine,trans-n,n'-dimethylcyclohexane-1,2-diamine,trans-n,n'-dimethyl-1,2-cyclohexanediamine,trans-1,2-bis methylamino cyclohexane,trans-1r,2r-n,n'-bismethyl-1,2-cyclohexane diamine,1r,2r---1,2-bis methylamino cyclohexane
2733821
(1R,2R)-1-N,2-N-dimethylcyclohexane-1,2-diamine
CNC1CCCCC1NC
Color White to Yellow
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
Specific Rotation − 143.00
Specific Rotation Condition −143° (20°C c=1,CHCl3)
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Linear Formula C6H10(NHCH3)2
Formula Weight 142.24
Physical Form Low Melting Solid
Percent Purity 95%
Chemical Name or Material (1R,2R)-N,N'-Dimethyl-1,2-cyclohexanediamine
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger