Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,2S)-1-Phenyl-2-(1-pyrrolidinyl)propan-1-ol 98.0+%, TCI America™

Click to view available options
Quantity:
1g
5g
127641-25-2
C13H19NO
205.301
MFCD01632702
FZVHJGJBJLFWEX-AAEUAGOBSA-N
(1R,2S)-N,N-Tetramethylenenorephedrine
10856631
(1R,2S)-1-phenyl-2-pyrrolidin-1-ylpropan-1-ol
CC(C(C1=CC=CC=C1)O)N2CCCC2
127641-25-2
C13H19NO
205.301
MFCD01632702
FZVHJGJBJLFWEX-AAEUAGOBSA-N
(1R,2S)-N,N-Tetramethylenenorephedrine
10856631
(1R,2S)-1-phenyl-2-pyrrolidin-1-ylpropan-1-ol
CC(C(C1=CC=CC=C1)O)N2CCCC2
Color White-Yellow
Quantity 1g
Formula Weight 205.30
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material (1R,2S)-1-Phenyl-2-(1-pyrrolidinyl)propan-1-ol