Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,2S)-(+)-2-Aminocyclohex-3-enecarboxylic acid hydrochloride, Acros Organics

Click to view available options
Quantity:
50mg
131783-54-5
C7H12ClNO2
177.628
UQGIRSZIKGUXTO-IBTYICNHSA-N
cis-2-amino-cyclohex-3-enecarboxylic acid hydrochloride,1r,2s-2-amino-cyclohex-3-enecarboxylic acid hydrochloride,cis-2-amino-3-cyclohexene-1-carboxylic acid hydrochloride,1r,2s-2-aminocyclohex-3-enecarboxylic acid hydrochloride,1r,2s-2-aminocyclohex-3-ene-1-carboxylic acid hydrochloride,2s,1r-2-aminocyclohex-3-enecarboxylic acid, chloride,1r,2s-2-amino-cyclohex-3-ene carboxylic acid hydrochloride,1r,2s-+-2-aminocyclohex-3-enecarboxylic acid hydrochloride
2734695
(1R,2S)-2-aminocyclohex-3-ene-1-carboxylic acid;hydrochloride
C1CC(C(C=C1)N)C(=O)O.Cl
131783-54-5
C7H12ClNO2
177.628
UQGIRSZIKGUXTO-IBTYICNHSA-N
cis-2-amino-cyclohex-3-enecarboxylic acid hydrochloride,1r,2s-2-amino-cyclohex-3-enecarboxylic acid hydrochloride,cis-2-amino-3-cyclohexene-1-carboxylic acid hydrochloride,1r,2s-2-aminocyclohex-3-enecarboxylic acid hydrochloride,1r,2s-2-aminocyclohex-3-ene-1-carboxylic acid hydrochloride,2s,1r-2-aminocyclohex-3-enecarboxylic acid, chloride,1r,2s-2-amino-cyclohex-3-ene carboxylic acid hydrochloride,1r,2s-+-2-aminocyclohex-3-enecarboxylic acid hydrochloride
2734695
(1R,2S)-2-aminocyclohex-3-ene-1-carboxylic acid;hydrochloride
C1CC(C(C=C1)N)C(=O)O.Cl
Quantity 50mg
Formula Weight 177.63
Chemical Name or Material (1R, 2S)-(+)-2-Aminocyclohex-3-enecarboxylic acid hydrochloride