Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,2S)-(-)-2-Aminocyclohexanecarboxylic acid hydrochloride, ACROS Organics™

Click to view available options
Quantity:
50mg
Packaging:
Glass bottle
158414-48-3
C7H14ClNO2
179.644
MFCD04037994
QNIOBHOEZYKCJV-IBTYICNHSA-N
1r,2s-2-aminocyclohexanecarboxylic acid hydrochloride,1r,2s---2-aminocyclohexanecarboxylic acid hydrochloride,1r,2s-2-aminocyclohexane-1-carboxylic acid hydrochloride,cis-2-aminocyclohexanecarboxylic acid hydrochloride,2s,1r-2-aminocyclohexanecarboxylic acid, chloride,cyclohexanecarboxylic acid, 2-amino-, hydrochloride, 1r,2s,cyclohexanecarboxylicacid,2-amino-,hydrochloride, 1r,2s-rel,cyclohexanecarboxylic acid, 2-amino-, hydrochloride, 1r,2s-rel
2734691
(1R,2S)-2-aminocyclohexane-1-carboxylic acid;hydrochloride
C1CCC(C(C1)C(=O)O)N.Cl
158414-48-3
C7H14ClNO2
179.644
MFCD04037994
QNIOBHOEZYKCJV-IBTYICNHSA-N
1r,2s-2-aminocyclohexanecarboxylic acid hydrochloride,1r,2s---2-aminocyclohexanecarboxylic acid hydrochloride,1r,2s-2-aminocyclohexane-1-carboxylic acid hydrochloride,cis-2-aminocyclohexanecarboxylic acid hydrochloride,2s,1r-2-aminocyclohexanecarboxylic acid, chloride,cyclohexanecarboxylic acid, 2-amino-, hydrochloride, 1r,2s,cyclohexanecarboxylicacid,2-amino-,hydrochloride, 1r,2s-rel,cyclohexanecarboxylic acid, 2-amino-, hydrochloride, 1r,2s-rel
2734691
(1R,2S)-2-aminocyclohexane-1-carboxylic acid;hydrochloride
C1CCC(C(C1)C(=O)O)N.Cl
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 50mg
Color Beige
Melting Point 234.0°C to 236.0°C
Formula Weight 179.69
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material (1R,2S)-(-)-2-Aminocyclohexanecarboxylic acid hydrochloride

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger