Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,2S,4S)-N-BOC-1-Amino-2-hydroxycyclopentane-4-carboxylic acid methyl ester, 95%, 98% e.e., ACROS Organics™

Click to view available options
Quantity:
250mg
500mg
Packaging:
Glass Bottle in Plastic Container
Glass bottle
321744-14-3
C12H21NO5
259.302
MFCD02259718
SSATZBUCOTXXGA-YIZRAAEISA-N
tert-butyl 1r,2s,4s-4-methoxycarbonyl-2-hydroxycyclopentyl-carbamate,1s,3r,4s-methyl 3-tert-butoxycarbonyl amino-4-hydroxycyclopentanecarboxylate,methyl 1s,3r,4s-3-tert-butoxycarbonyl amino-4-hydroxycyclopentane-1-carboxylate,tert-butyl 1r,2s,4s-4-methoxycarbonyl-2-hydroxycyclopentylcarbamate,1r,2s,4s-n-boc-1-amino-2-hydroxycyclopentane-4-carboxylic acid methyl ester,1s,3r,4s-3-tert-butoxycarbonylamino-4-hydroxycyclopentanecarboxylic acid methyl ester,methyl 1s,3r,4s-3-tert-butoxy carbonyl amino-4-hydroxycyclopentane-1-carboxylate,n-boc-1 r-amino-2 s-hydroxy-cyclopentane-4 s-carboxylic acid methyl ester,3beta-tert-butyloxycarbonyl amino-4beta-hydroxycyclopentane-1beta-carboxylic acid methyl ester,cyclopentanecarboxylic acid, 3-1,1-dimethylethoxy carbonyl amino-4-hydroxy-, methyl ester, 1s,3r,4s-9ci
2734526
methyl (1S,3S,4R)-3-hydroxy-4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylate
CC(C)(C)OC(=O)NC1CC(CC1O)C(=O)OC
321744-14-3
C12H21NO5
259.302
MFCD02259718
SSATZBUCOTXXGA-YIZRAAEISA-N
tert-butyl 1r,2s,4s-4-methoxycarbonyl-2-hydroxycyclopentyl-carbamate,1s,3r,4s-methyl 3-tert-butoxycarbonyl amino-4-hydroxycyclopentanecarboxylate,methyl 1s,3r,4s-3-tert-butoxycarbonyl amino-4-hydroxycyclopentane-1-carboxylate,tert-butyl 1r,2s,4s-4-methoxycarbonyl-2-hydroxycyclopentylcarbamate,1r,2s,4s-n-boc-1-amino-2-hydroxycyclopentane-4-carboxylic acid methyl ester,1s,3r,4s-3-tert-butoxycarbonylamino-4-hydroxycyclopentanecarboxylic acid methyl ester,methyl 1s,3r,4s-3-tert-butoxy carbonyl amino-4-hydroxycyclopentane-1-carboxylate,n-boc-1 r-amino-2 s-hydroxy-cyclopentane-4 s-carboxylic acid methyl ester,3beta-tert-butyloxycarbonyl amino-4beta-hydroxycyclopentane-1beta-carboxylic acid methyl ester,cyclopentanecarboxylic acid, 3-1,1-dimethylethoxy carbonyl amino-4-hydroxy-, methyl ester, 1s,3r,4s-9ci
2734526
methyl (1S,3S,4R)-3-hydroxy-4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylate
CC(C)(C)OC(=O)NC1CC(CC1O)C(=O)OC
Enantiomeric Excess 97.5% min. (GC)
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 250mg
Color Colorless to Yellow
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 259.3
Physical Form Viscous Liquid
Percent Purity ≥94%
Chemical Name or Material (1R, 2S, 4S)-N-BOC-1-Amino-2-hydroxycyclopentane-4-carboxylic acid methyl ester, 95%, (98% e.e.)
Show More Show Less