Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,2S)-(+)-cis-2-(Benzylamino)cyclohexanemethanol, 99%, 99% e.e., ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
71581-92-5
C14H21NO
219.33
MFCD00151395
BRQFIORUNWWNBM-KBPBESRZSA-N
1r,2s-2-benzylamino cyclohexyl methanol,+-cis-2-benzylaminocyclohexanemethanol,cis-1r,2s-+-2-benzylamino cyclohexanemethanol,--cis-2-benzylaminocyclohexanemethanol,cis-1r,2s-+-2-benzylaminocyclohexanemethanol,pubchem7939,cis-2-benzylamino cyclohexylmethanol,2-benzylamino cyclohexyl methanol #,cis-2-benzylamino cyclohexyl methanol
853687
[(1R,2S)-2-(benzylamino)cyclohexyl]methanol
C1CCC(C(C1)CO)NCC2=CC=CC=C2
71581-92-5
C14H21NO
219.33
MFCD00151395
BRQFIORUNWWNBM-KBPBESRZSA-N
1r,2s-2-benzylamino cyclohexyl methanol,+-cis-2-benzylaminocyclohexanemethanol,cis-1r,2s-+-2-benzylamino cyclohexanemethanol,--cis-2-benzylaminocyclohexanemethanol,cis-1r,2s-+-2-benzylaminocyclohexanemethanol,pubchem7939,cis-2-benzylamino cyclohexylmethanol,2-benzylamino cyclohexyl methanol #,cis-2-benzylamino cyclohexyl methanol
853687
[(1R,2S)-2-(benzylamino)cyclohexyl]methanol
C1CCC(C(C1)CO)NCC2=CC=CC=C2
Melting Point 66°C to 69°C
Packaging Glass bottle
Color White
Infrared Spectrum Authentic
Quantity 5g
Enantiomeric Excess 98.5% min.
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula C6H5CH2NHC6H10CH2OH
Formula Weight 219.33
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material (1R, 2S)-(+)-cis-2-(Benzylamino)cyclohexanemethanol, 99% e.e.
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.