Alfa Aesar™ (1R,3S)-(-)-3-Aminocyclopentanecarboxylic acid, 95%

Manufacturer: Alfa Aesar™ H52122MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH52122MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 71830-08-5
    Molecular Formula C6H11NO2
    Formula Weight 129.16
    MDL Number MFCD00211289
    Chemical Name or Material (1R,3S)-(-)-3-Aminocyclopentanecarboxylic acid

    Specifications

    Formula Weight 129.16
    Molecular Formula C6H11NO2
    MDL Number MFCD00211289
    CAS 71830-08-5
    Quantity 250mg
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material (1R,3S)-(-)-3-Aminocyclopentanecarboxylic acid
    Assay 95%

    Description

    Applications
    (1R,3S)-(-)-3-Aminocyclopentanecarboxylic acid is used as pharmaceutical intermediate.

    Solubility
    Slightly soluble in water.

    Notes
    Air sensitive. Incompatible with oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit