Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,3S)-3-Aminocyclopentanecarboxylic acid, 95%, 98% ee, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
71830-08-5
C6H11NO2
129.16
MLLSSTJTARJLHK-UHNVWZDZSA-N
1r,3s-3-aminocyclopentanecarboxylic acid,1r,3s-3-aminocyclopentane-1-carboxylic acid,--1r,3s-3-aminocyclopentanecarboxylic acid,cis-3-aminocyclopentanecarboxylic acid,cyclopentanecarboxylic acid, 3-amino-, 1r,3s,1r,3s---3-aminocyclopentanecarboxylic acid,cis-3-amino-cyclopentanecarboxylic acid,1r,3s-3-amino-cyclopentanecarboxylic acid,pubchem18731,1r,3s---3-amino-cyclopentanecarboxylic acid
1502035
(1R,3S)-3-aminocyclopentane-1-carboxylic acid
C1CC(CC1C(=O)O)N
71830-08-5
C6H11NO2
129.16
MLLSSTJTARJLHK-UHNVWZDZSA-N
1r,3s-3-aminocyclopentanecarboxylic acid,1r,3s-3-aminocyclopentane-1-carboxylic acid,--1r,3s-3-aminocyclopentanecarboxylic acid,cis-3-aminocyclopentanecarboxylic acid,cyclopentanecarboxylic acid, 3-amino-, 1r,3s,1r,3s---3-aminocyclopentanecarboxylic acid,cis-3-amino-cyclopentanecarboxylic acid,1r,3s-3-amino-cyclopentanecarboxylic acid,pubchem18731,1r,3s---3-amino-cyclopentanecarboxylic acid
1502035
(1R,3S)-3-aminocyclopentane-1-carboxylic acid
C1CC(CC1C(=O)O)N
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
Color White to Beige
Enantiomeric Excess 97.5% (HPLC) min.
Melting Point 172.1°C
Assay Percent Range 94% min. (HPLC)
Formula Weight 129.16
Physical Form Crystalline Powder
Percent Purity 95%,98% ee
Chemical Name or Material (1R, 3S)-3-Aminocyclopentanecarboxylic acid
Show More Show Less

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

GHS Signal Word: Warning