Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,4R)-4-(BOC-Amino)cyclopent-2-enecarboxylic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
298716-03-7
C11H17NO4
227.26
WOUNTSATDZJBLP-YUMQZZPRSA-N
1r,4r-4-boc-amino cyclopent-2-enecarboxylic acid,1r,4r-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid,1r,4r-4-tert-butoxycarbonylamino cyclopent-2-enecarboxylic acid,1r,4r-4-tert-butoxycarbonyl amino cyclopent-2-ene-1-carboxylic acid,pubchem17360,1s,4r---4-tert-butoxycarbonyl aminocyclopent-2-ene-1-carboxylic acid,trans-4-boc-amino-2-cyclopentene-1-carboxylic acid,1r,4r-4-boc-amino cyclopent-2-enecarboxylicacid,trans-1r,4r-4-boc-amino-2-cyclopentene-1-carboxylic acid
7168126
(1R,4R)-4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopent-2-ene-1-carboxylic acid
CC(C)(C)OC(=O)NC1CC(C=C1)C(=O)O
298716-03-7
C11H17NO4
227.26
WOUNTSATDZJBLP-YUMQZZPRSA-N
1r,4r-4-boc-amino cyclopent-2-enecarboxylic acid,1r,4r-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid,1r,4r-4-tert-butoxycarbonylamino cyclopent-2-enecarboxylic acid,1r,4r-4-tert-butoxycarbonyl amino cyclopent-2-ene-1-carboxylic acid,pubchem17360,1s,4r---4-tert-butoxycarbonyl aminocyclopent-2-ene-1-carboxylic acid,trans-4-boc-amino-2-cyclopentene-1-carboxylic acid,1r,4r-4-boc-amino cyclopent-2-enecarboxylicacid,trans-1r,4r-4-boc-amino-2-cyclopentene-1-carboxylic acid
7168126
(1R,4R)-4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopent-2-ene-1-carboxylic acid
CC(C)(C)OC(=O)NC1CC(C=C1)C(=O)O
Quantity 1g
Melting Point 110.8°C to 113.1°C
Color White
Infrared Spectrum Authentic
Specific Rotation +240° to +260° (20°C, 589nm) c=0.5, methanol
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 227.26
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material (1R, 4R)-4-(BOC-Amino)cyclopent-2-enecarboxylic acid
Show More Show Less