Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,4S)-4-Aminocyclopent-2-enecarboxylic acid, 95%, 98% ee, Acros Organics

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
134003-04-6
C6H9NO2
127.14
MFCD00211285
VTCHZFWYUPZZKL-CRCLSJGQSA-N
1r,4s-4-aminocyclopent-2-enecarboxylic acid,1r,4s-4-aminocyclopent-2-ene-1-carboxylic acid,1r,4s-rel-4-aminocyclopent-2-enecarboxylic acid,+-1r,4s-4-aminocyclopent-2-enecarboxylic acid,1r,4s-4-amino-2-cyclopentene-1-carboxylic acid,1r,4s-4-amino-cyclopent-2-ene carboxylic acid,1r,4s-+-4-aminocyclopent-2-enecarboxylic acid,2-cyclopentene-1-carboxylic acid, 4-amino-, 1r,4s,pubchem18728,+-1r,4s-g-homocycloleu-2-ene
2734525
(1R,4S)-4-aminocyclopent-2-ene-1-carboxylic acid
C1C(C=CC1N)C(=O)O
134003-04-6
C6H9NO2
127.14
MFCD00211285
VTCHZFWYUPZZKL-CRCLSJGQSA-N
1r,4s-4-aminocyclopent-2-enecarboxylic acid,1r,4s-4-aminocyclopent-2-ene-1-carboxylic acid,1r,4s-rel-4-aminocyclopent-2-enecarboxylic acid,+-1r,4s-4-aminocyclopent-2-enecarboxylic acid,1r,4s-4-amino-2-cyclopentene-1-carboxylic acid,1r,4s-4-amino-cyclopent-2-ene carboxylic acid,1r,4s-+-4-aminocyclopent-2-enecarboxylic acid,2-cyclopentene-1-carboxylic acid, 4-amino-, 1r,4s,pubchem18728,+-1r,4s-g-homocycloleu-2-ene
2734525
(1R,4S)-4-aminocyclopent-2-ene-1-carboxylic acid
C1C(C=CC1N)C(=O)O
Melting Point 180°C
Quantity 1g
Color Brown
Packaging Glass bottle
Infrared Spectrum Authentic
Enantiomeric Excess 97.5% min. (HPLC)
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 127.14
Physical Form Crystalline Powder or Flakes
Percent Purity ≥94%
Chemical Name or Material (1R, 4S)-4-Aminocyclopent-2-enecarboxylic acid, 95%, (98% e.e.)
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning